Anti-TMPRSS5 Mouse Monoclonal Antibody [clone: 376608]

Supplier: R & D Systems
MAB2495
220019-893EA 763.57 USD
220019-893
Anti-TMPRSS5 Mouse Monoclonal Antibody [clone: 376608]
Antibodies
Type: Primary
Antigen: TMPRSS5
Clonality: Monoclonal
Clone: 376608
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR