Anti-XPNPEP2 Mouse Monoclonal Antibody [clone: 391131]

Supplier: R & D Systems
MAB24901
220019-892EA 561.11 USD
220019-892
Anti-XPNPEP2 Mouse Monoclonal Antibody [clone: 391131]
Antibodies
Type: Primary
Antigen: XPNPEP2
Clonality: Monoclonal
Clone: 391131
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR