Anti-CNDP1 Mouse Monoclonal Antibody [clone: 352216]

Supplier: R & D Systems
MAB2489
220019-888EA 594.37 USD
220019-888
Anti-CNDP1 Mouse Monoclonal Antibody [clone: 352216]
Antibodies
Type: Primary
Antigen: CNDP1
Clonality: Monoclonal
Clone: 352216
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR