Anti-SERPING1 Mouse Monoclonal Antibody [clone: 350507]

Supplier: R & D Systems
MAB2488
220019-887EA 623.29 USD
220019-887
Anti-SERPING1 Mouse Monoclonal Antibody [clone: 350507]
Antibodies
Type: Primary
Antigen: SERPING1
Clonality: Monoclonal
Clone: 350507
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR