Anti-ARSA Mouse Monoclonal Antibody [clone: 324802]

Supplier: R & D Systems
MAB2485
220019-885EA 594.37 USD
220019-885
Anti-ARSA Mouse Monoclonal Antibody [clone: 324802]
Antibodies
Type: Primary
Antigen: ARSA
Clonality: Monoclonal
Clone: 324802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR