Anti-PSPN Rat Monoclonal Antibody [clone: 240839]

Supplier: R & D Systems
MAB2479
220019-882EA 763.57 USD
220019-882
Anti-PSPN Rat Monoclonal Antibody [clone: 240839]
Antibodies
Type: Primary
Antigen: PSPN
Clonality: Monoclonal
Clone: 240839
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR