Anti-BRCA2 Mouse Monoclonal Antibody [clone: 234403]

Supplier: R & D Systems
MAB2476
220019-881EA 639.2 USD
220019-881
Anti-BRCA2 Mouse Monoclonal Antibody [clone: 234403]
Antibodies
Type: Primary
Antigen: BRCA2
Clonality: Monoclonal
Clone: 234403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR