Anti-IL4 Mouse Monoclonal Antibody [clone: 701214]

Supplier: R & D Systems
MAB2469
102885-464EA 577.02 USD
102885-464
Anti-IL4 Mouse Monoclonal Antibody [clone: 701214]
Antibodies
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 701214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR