Anti-TGFB1 Mouse Monoclonal Antibody [clone: 27232]

Supplier: R & D Systems
MAB2463
220019-877EA 763.57 USD
220019-877
Anti-TGFB1 Mouse Monoclonal Antibody [clone: 27232]
Antibodies
Type: Primary
Antigen: TGFB1
Clonality: Monoclonal
Clone: 27232
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR