Anti-F11 Mouse Monoclonal Antibody [clone: 318104]

Supplier: R & D Systems
MAB24601
220019-875EA 772.25 USD
220019-875
Anti-F11 Mouse Monoclonal Antibody [clone: 318104]
Antibodies
Type: Primary
Antigen: F11
Clonality: Monoclonal
Clone: 318104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR