Anti-F11 Mouse Monoclonal Antibody [clone: 318101]

Supplier: R & D Systems
MAB2460
220019-874EA 594.37 USD
220019-874
Anti-F11 Mouse Monoclonal Antibody [clone: 318101]
Antibodies
Type: Primary
Antigen: F11
Clonality: Monoclonal
Clone: 318101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR