Anti-OLIG3 Mouse Monoclonal Antibody [clone: 257934]

Supplier: R & D Systems
MAB2456
220019-870EA 594.37 USD
220019-870
Anti-OLIG3 Mouse Monoclonal Antibody [clone: 257934]
Antibodies
Type: Primary
Antigen: OLIG3
Clonality: Monoclonal
Clone: 257934
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR