Anti-PRSS50 Rat Monoclonal Antibody [clone: 260514]

Supplier: R & D Systems
MAB2455
220019-869EA 772.25 USD
220019-869
Anti-PRSS50 Rat Monoclonal Antibody [clone: 260514]
Antibodies
Type: Primary
Antigen: PRSS50
Clonality: Monoclonal
Clone: 260514
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR