Anti-ROBO4 Mouse Monoclonal Antibody [clone: 265721]

Supplier: R & D Systems
MAB24541
220019-868EA 561.11 USD
220019-868
Anti-ROBO4 Mouse Monoclonal Antibody [clone: 265721]
Antibodies
Type: Primary
Antigen: ROBO4
Clonality: Monoclonal
Clone: 265721
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR