Anti-ROBO4 Mouse Monoclonal Antibody [clone: 265703]

Supplier: R & D Systems
MAB2454
220019-867EA 577.02 USD
220019-867
Anti-ROBO4 Mouse Monoclonal Antibody [clone: 265703]
Antibodies
Type: Primary
Antigen: ROBO4
Clonality: Monoclonal
Clone: 265703
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR