Anti-IGF2R Mouse Monoclonal Antibody [clone: 372616]

Supplier: R & D Systems
MAB24471
220019-860EA 772.25 USD
220019-860
Anti-IGF2R Mouse Monoclonal Antibody [clone: 372616]
Antibodies
Type: Primary
Antigen: IGF2R
Clonality: Monoclonal
Clone: 372616
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR