Anti-IGF2R Mouse Monoclonal Antibody [clone: 372604]

Supplier: R & D Systems
MAB2447
220019-859EA 594.37 USD
220019-859
Anti-IGF2R Mouse Monoclonal Antibody [clone: 372604]
Antibodies
Type: Primary
Antigen: IGF2R
Clonality: Monoclonal
Clone: 372604
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR