Anti-CHIL3 Rat Monoclonal Antibody [clone: 281926]

Supplier: R & D Systems
MAB2446
220019-858EA 594.37 USD
220019-858
Anti-CHIL3 Rat Monoclonal Antibody [clone: 281926]
Antibodies
Type: Primary
Antigen: CHIL3
Clonality: Monoclonal
Clone: 281926
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR