Anti-NKX2-5 Mouse Monoclonal Antibody [clone: 259416]

Supplier: R & D Systems
MAB2444
220019-856EA 633.42 USD
220019-856
Anti-NKX2-5 Mouse Monoclonal Antibody [clone: 259416]
Antibodies
Type: Primary
Antigen Symbol: NKX2-5
Antigen Name: NK2 homeobox 5
Antigen Synonyms: CHNG5, CSX, CSX1, HLHS2, NKX2.5, NKX2E, NKX4-1, VSD3
Clonality: Monoclonal
Clone: 259416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR