Anti-FZD9 Rat Monoclonal Antibody [clone: 291004]

Supplier: R & D Systems
MAB2440
220019-855EA 561.11 USD
220019-855
Anti-FZD9 Rat Monoclonal Antibody [clone: 291004]
Antibodies
Type: Primary
Antigen Symbol: FZD9
Antigen Name: frizzled homolog 9 (Drosophila)
Antigen Synonyms: mfz9
Clonality: Monoclonal
Clone: 291004
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR