Anti-ICAM2 Mouse Monoclonal Antibody [clone: 86911]

Supplier: R & D Systems
MAB244
220019-854EA 763.57 USD
220019-854
Anti-ICAM2 Mouse Monoclonal Antibody [clone: 86911]
Antibodies
Type: Primary
Antigen: ICAM2
Clonality: Monoclonal
Clone: 86911
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR