Anti-MOG Rat Monoclonal Antibody [clone: 296829]

Supplier: R & D Systems
MAB2439
220019-853EA 600.15 USD
220019-853
Anti-MOG Rat Monoclonal Antibody [clone: 296829]
Antibodies
Type: Primary
Antigen: MOG
Clonality: Monoclonal
Clone: 296829
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR