Anti-IL2RA Rat Monoclonal Antibody [clone: 280406]

Supplier: R & D Systems
MAB2438
220019-852EA 594.37 USD
220019-852
Anti-IL2RA Rat Monoclonal Antibody [clone: 280406]
Antibodies
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 280406
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR