Anti-ADAM33 Rat Monoclonal Antibody [clone: 219515]

Supplier: R & D Systems
MAB2434
220019-851EA 561.11 USD
220019-851
Anti-ADAM33 Rat Monoclonal Antibody [clone: 219515]
Antibodies
Type: Primary
Antigen: ADAM33
Clonality: Monoclonal
Clone: 219515
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR