Anti-PRSS50 Rat Monoclonal Antibody [clone: 260606]

Supplier: R & D Systems
MAB2430
220019-850EA 561.11 USD
220019-850
Anti-PRSS50 Rat Monoclonal Antibody [clone: 260606]
Antibodies
Type: Primary
Antigen: PRSS50
Clonality: Monoclonal
Clone: 260606
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR