Anti-CDON Rat Monoclonal Antibody [clone: 274133]

Supplier: R & D Systems
MAB2429
220019-848EA 561.11 USD
220019-848
Anti-CDON Rat Monoclonal Antibody [clone: 274133]
Antibodies
Type: Primary
Antigen: CDON
Clonality: Monoclonal
Clone: 274133
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR