Anti-FCN2 Mouse Monoclonal Antibody [clone: 297018]

Supplier: R & D Systems
MAB2428
220019-847EA 577.02 USD
220019-847
Anti-FCN2 Mouse Monoclonal Antibody [clone: 297018]
Antibodies
Type: Primary
Antigen Symbol: FCN2
Antigen Name: ficolin (collagen/fibrinogen domain containing lectin) 2
Antigen Synonyms: EBP-37, FCNL, P35, ficolin-2
Clonality: Monoclonal
Clone: 297018
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR