Anti-IL1F10 Rat Monoclonal Antibody [clone: 316709]

Supplier: R & D Systems
MAB2427
220019-846EA 594.37 USD
220019-846
Anti-IL1F10 Rat Monoclonal Antibody [clone: 316709]
Antibodies
Type: Primary
Antigen: IL1F10
Clonality: Monoclonal
Clone: 316709
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR