Anti-FCRL4 Mouse Monoclonal Antibody [clone: 580810]

Supplier: R & D Systems
MAB24262
102885-462EA 594.37 USD
102885-462
Anti-FCRL4 Mouse Monoclonal Antibody [clone: 580810]
Antibodies
Type: Primary
Antigen Symbol: FCRL4
Antigen Name: Fc receptor-like 4
Antigen Synonyms: CD307d, FCRH4, IGFP2, IRTA1
Clonality: Monoclonal
Clone: 580810
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR