Anti-LILRB4 Mouse Monoclonal Antibody [clone: 293623]

Supplier: R & D Systems
MAB24251
220019-845EA 655.11 USD
220019-845
Anti-LILRB4 Mouse Monoclonal Antibody [clone: 293623]
Antibodies
Type: Primary
Antigen: LILRB4
Clonality: Monoclonal
Clone: 293623
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR