Anti-TREML1 Rat Monoclonal Antibody [clone: 268529]

Supplier: R & D Systems
MAB2424
220019-842EA 577.02 USD
220019-842
Anti-TREML1 Rat Monoclonal Antibody [clone: 268529]
Antibodies
Type: Primary
Antigen: TREML1
Clonality: Monoclonal
Clone: 268529
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR