Anti-GRN Mouse Monoclonal Antibody [clone: 296628]

Supplier: R & D Systems
MAB2420
220019-840EA 655.11 USD
220019-840
Anti-GRN Mouse Monoclonal Antibody [clone: 296628]
Antibodies
Type: Primary
Antigen Symbol: GRN
Antigen Name: granulin
Antigen Synonyms: CLN11, GEP, GP88, PCDGF, PEPI, PGRN
Clonality: Monoclonal
Clone: 296628
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR