Anti-TGFBR3 Mouse Monoclonal Antibody [clone: 31349]

Supplier: R & D Systems
MAB242
220019-839EA 772.25 USD
220019-839
Anti-TGFBR3 Mouse Monoclonal Antibody [clone: 31349]
Antibodies
Type: Primary
Antigen: TGFBR3
Clonality: Monoclonal
Clone: 31349
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR