Anti-PDX1 Mouse Monoclonal Antibody [clone: 267712]

Supplier: R & D Systems
MAB2419
220019-838EA 647.88 USD
220019-838
Anti-PDX1 Mouse Monoclonal Antibody [clone: 267712]
Antibodies
Type: Primary
Antigen: PDX1
Clonality: Monoclonal
Clone: 267712
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR