Anti-OLIG1 Mouse Monoclonal Antibody [clone: 257219]

Supplier: R & D Systems
MAB2417
220019-837EA 600.15 USD
220019-837
Anti-OLIG1 Mouse Monoclonal Antibody [clone: 257219]
Antibodies
Type: Primary
Antigen: OLIG1
Clonality: Monoclonal
Clone: 257219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR