Anti-SCARF1 Mouse Monoclonal Antibody [clone: 373606]

Supplier: R & D Systems
MAB2409
220019-834EA 594.37 USD
220019-834
Anti-SCARF1 Mouse Monoclonal Antibody [clone: 373606]
Antibodies
Type: Primary
Antigen: SCARF1
Clonality: Monoclonal
Clone: 373606
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR