Anti-NCAM1 Mouse Monoclonal Antibody [clone: 301040]

Supplier: R & D Systems
MAB2408
220019-832EA 772.25 USD
220019-832
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 301040]
Antibodies
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 301040
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR