Anti-BMPR1A Mouse Monoclonal Antibody [clone: 550607]

Supplier: R & D Systems
MAB24061
220019-831EA 594.37 USD
220019-831
Anti-BMPR1A Mouse Monoclonal Antibody [clone: 550607]
Antibodies
Type: Primary
Antigen: BMPR1A
Clonality: Monoclonal
Clone: 550607
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR