Anti-ITGB1 Rat Monoclonal Antibody [clone: 265917]

Supplier: R & D Systems
MAB2405
220019-829EA 633.42 USD
220019-829
Anti-ITGB1 Rat Monoclonal Antibody [clone: 265917]
Antibodies
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: 265917
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR