Anti-CD38 Mouse Monoclonal Antibody [clone: 240726]

Supplier: R & D Systems
MAB24041
220019-828EA 594.37 USD
220019-828
Anti-CD38 Mouse Monoclonal Antibody [clone: 240726]
Antibodies
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 240726
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR