Anti-RUNX1 Mouse Monoclonal Antibody [clone: 751926]

Supplier: R & D Systems
MAB23991
102885-458EA 594.37 USD
102885-458
Anti-RUNX1 Mouse Monoclonal Antibody [clone: 751926]
Antibodies
Type: Primary
Antigen: RUNX1
Clonality: Monoclonal
Clone: 751926
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR