Anti-CRISP3 Mouse Monoclonal Antibody [clone: 295208]

Supplier: R & D Systems
MAB23972
220019-823EA 561.11 USD
220019-823
Anti-CRISP3 Mouse Monoclonal Antibody [clone: 295208]
Antibodies
Type: Primary
Antigen: CRISP3
Clonality: Monoclonal
Clone: 295208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR