Anti-CRISP3 Mouse Monoclonal Antibody [clone: 295203]

Supplier: R & D Systems
MAB23971
220019-822EA 577.02 USD
220019-822
Anti-CRISP3 Mouse Monoclonal Antibody [clone: 295203]
Antibodies
Type: Primary
Antigen: CRISP3
Clonality: Monoclonal
Clone: 295203
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR