Anti-DDR1 Mouse Monoclonal Antibody [clone: 290420]

Supplier: R & D Systems
MAB2396
220019-820EA 594.37 USD
220019-820
Anti-DDR1 Mouse Monoclonal Antibody [clone: 290420]
Antibodies
Type: Primary
Antigen: DDR1
Clonality: Monoclonal
Clone: 290420
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR