Anti-TREML1 Rat Monoclonal Antibody [clone: 268420]

Supplier: R & D Systems
MAB2394
220019-819EA 594.37 USD
220019-819
Anti-TREML1 Rat Monoclonal Antibody [clone: 268420]
Antibodies
Type: Primary
Antigen: TREML1
Clonality: Monoclonal
Clone: 268420
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR