Anti-THBS4 Mouse Monoclonal Antibody [clone: 276523]

Supplier: R & D Systems
MAB2390
220019-818EA 594.37 USD
220019-818
Anti-THBS4 Mouse Monoclonal Antibody [clone: 276523]
Antibodies
Type: Primary
Antigen: THBS4
Clonality: Monoclonal
Clone: 276523
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR