Anti-ITGB6 Rat Monoclonal Antibody [clone: 268622]

Supplier: R & D Systems
MAB2389
220019-817EA 594.37 USD
220019-817
Anti-ITGB6 Rat Monoclonal Antibody [clone: 268622]
Antibodies
Type: Primary
Antigen: ITGB6
Clonality: Monoclonal
Clone: 268622
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR