Anti-CD69 Rat Monoclonal Antibody [clone: 310116]

Supplier: R & D Systems
MAB23861
220019-815EA 594.37 USD
220019-815
Anti-CD69 Rat Monoclonal Antibody [clone: 310116]
Antibodies
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: 310116
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR