Anti-CLEC5A Mouse Monoclonal Antibody [clone: 283834]

Supplier: R & D Systems
MAB2384
220019-813EA 655.11 USD
220019-813
Anti-CLEC5A Mouse Monoclonal Antibody [clone: 283834]
Antibodies
Type: Primary
Antigen: CLEC5A
Clonality: Monoclonal
Clone: 283834
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR