Anti-CREG1 Mouse Monoclonal Antibody [clone: 299133]

Supplier: R & D Systems
MAB2380
220019-812EA 577.02 USD
220019-812
Anti-CREG1 Mouse Monoclonal Antibody [clone: 299133]
Antibodies
Type: Primary
Antigen: CREG1
Clonality: Monoclonal
Clone: 299133
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR